Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 96-well & 384-well Microtiter Plates, Holds 27 with Lids, 36 without Lids, 10 15/16 x 7 5/16 x 5 7/16"
SDP
Catalog No. 503027824
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks for 96-Well & 384-Well Microtiter Plates, 96-Well Microtube Boxes and Micronics ROBO Racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders.  Drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Plate Capacity: 27 with lids 36 without lids 3×3 Configuration. Dimensions (DxWxH): 11 1/8 x 5 7/16 x 7 5/16 inches (282 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-7824 Supplier Alkali Scientific Supplier No. UFDMP33
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks for 96-Well & 384-Well Microtiter Plates, 96-Well Microtube Boxes and Micronics ROBO Racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders.  Drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Plate Capacity: 27 with lids 36 without lids 3×3 Configuration. Dimensions (DxWxH): 11 1/8 x 5 7/16 x 7 5/16 inches (282 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.