Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 8ml, 8.5ml, 10ml Tubes, 60 Capacity, 1 Drawer
SDP
Catalog No. 503028396
Encompass_Preferred
For Blood Sample Tubes8ml, 8.5ml, 10ml Tube Capacity: 60 Tubes1 Drawer Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 60 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 4 1/2 inches (424 x 140 x 114mm)
Catalog No. 50-302-8396 Supplier Alkali Scientific Supplier No. UFDRT101
For Blood Sample Tubes8ml, 8.5ml, 10ml Tube Capacity: 60 Tubes1 Drawer Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 60 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 4 1/2 inches (424 x 140 x 114mm)