Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 3mL Tubes, 180 Capacity, 2 Drawers
SDP
Catalog No. 503027791
Encompass_Preferred
Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 180 tubes for 3ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 6 1/16 inches (424 x 140 x 154mm)
Catalog No. 50-302-7791 Supplier Alkali Scientific Supplier No. UFDRT032
Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 180 tubes for 3ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 6 1/16 inches (424 x 140 x 154mm)