Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 1mL Tubes, 360 Capacity, 3 Drawers
SDP
Catalog No. 503027793
Encompass_Preferred
Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 360 tubes for 1ml sizes. This freezer rack features three drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 9 1/8 inches (424 x 140 x 231mm)
Catalog No. 50-302-7793 Supplier Alkali Scientific Supplier No. UFDRT013
Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 360 tubes for 1ml sizes. This freezer rack features three drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 9 1/8 inches (424 x 140 x 231mm)