Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 50ml Centrifuge Tubes – Rack Only (1 Drawer, 51 Tubes)
SDP
Catalog No. 503027828
Encompass_Preferred
For 50ml Centrifuge Tubes Upright Freezer Drawer Rack stores 50 ml Centrifuge Tubes and features 1 drawer for storage up to 51 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer. Dimensions (DxWxH): 26 15/16  x  4 15/16 x 5 1/2  inches (685 x 126 x 140mm)Capacity: 51 Tubes *Lock device is ordered separately.
Catalog No. 50-302-7828 Supplier Alkali Scientific Supplier No. UFDRXLT501
For 50ml Centrifuge Tubes Upright Freezer Drawer Rack stores 50 ml Centrifuge Tubes and features 1 drawer for storage up to 51 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer. Dimensions (DxWxH): 26 15/16  x  4 15/16 x 5 1/2  inches (685 x 126 x 140mm)Capacity: 51 Tubes *Lock device is ordered separately.