Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 15ml Centrifuge Tubes – Rack Only (1 Drawer; 80 Tubes)
SDP
Catalog No. 503027819
Encompass_Preferred
For 15ml Centrifuge Tubes Upright Freezer Drawer Rack stores 15 ml Centrifuge Tubes and features 1 drawer for storage up to 80 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer.  Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 15/16 inches (564 x 140 x 126mm)Capacity: 80 Tubes
Catalog No. 50-302-7819 Supplier Alkali Scientific Supplier No. UFDRLT151
For 15ml Centrifuge Tubes Upright Freezer Drawer Rack stores 15 ml Centrifuge Tubes and features 1 drawer for storage up to 80 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer.  Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 15/16 inches (564 x 140 x 126mm)Capacity: 80 Tubes