Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 8ml, 8.5ml, 10ml Tubes, 120 Capacity, 2 Drawers
SDP
Catalog No. 503028566
Encompass_Preferred
For Blood Sample Tubes8ml, 8.5ml, 10ml Tube Capacity: 120 Tubes2 Drawers Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 120 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 9 inches (424 x 140 x 228mm)Bottom of Form
Catalog No. 50-302-8566 Supplier Alkali Scientific Supplier No. UFDRT102
For Blood Sample Tubes8ml, 8.5ml, 10ml Tube Capacity: 120 Tubes2 Drawers Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 120 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 9 inches (424 x 140 x 228mm)Bottom of Form