Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 96-well & 384-well Microtiter Plates, Holds 45 with Lids, 60 without Lids, 18 x 7 5/16 x 5 7/16"
SDP
Catalog No. 503029026
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks for 96-Well & 384-Well Microtiter Plates, 96-Well Microtube Boxes and Micronic ROBO Racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders.  Drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up. Plate Capacity: 45 with lids 60 without lids 5×3 Configuration Dimensions (DxWxH): 18 3/16 x 5 7/16 x 7 5/16 inches (462 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-9026 Supplier Alkali Scientific Supplier No. UFDMP53
For 96-Well and 384-Well Microtiter Plates For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks for 96-Well & 384-Well Microtiter Plates, 96-Well Microtube Boxes and Micronic ROBO Racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders.  Drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up. Plate Capacity: 45 with lids 60 without lids 5×3 Configuration Dimensions (DxWxH): 18 3/16 x 5 7/16 x 7 5/16 inches (462 x 138 x 186mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.