Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Microtiter Plates, 7 x 5 configuration, 35 Box Capacity
SDP
Catalog No. 503029028
Encompass_Preferred
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 35 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Security lock devices are also available for each rack though ordered separately. Plate Capacity: 105 with lids 140 without lids7×5 Configuration Dimensions (DxWxH): 25 1/4 x 5 7/16 x 12 1/8 inches (642 x 138 x 308mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-9028 Supplier Alkali Scientific Supplier No. UFDMP75
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm) Upright Freezer Drawer Racks allow for storage of 35 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.  Security lock devices are also available for each rack though ordered separately. Plate Capacity: 105 with lids 140 without lids7×5 Configuration Dimensions (DxWxH): 25 1/4 x 5 7/16 x 12 1/8 inches (642 x 138 x 308mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.