Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 96-well & 384-well Microtiter Plates, Holds 48 with Lids, 64 without Lids, 14 1/2 x 9 11/16 x 5 7/16"
SDP
Catalog No. 503029040
Encompass_Preferred
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm)  Upright Freezer Drawer Racks allow for storage of 16 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up  Plate Capacity: 48 with lids 64 without lids 4×4 Configuration Dimensions (DxWxH): 14 5/8 x 5 7/16 x 9 3/4 inches (372 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.
Catalog No. 50-302-9040 Supplier Alkali Scientific Supplier No. UFDMP44
For 96-Well and 384-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene) H: 11/16″-2 1/8″ (17.3-53.5mm)  Upright Freezer Drawer Racks allow for storage of 16 total 96/384-Well Microtiter Plates, Microtube Boxes or Micronic ROBO Racks. These freezer racks are constructed of corrosion resistant stainless steel.  Racks include handles and identification card holders for easy access and identification.  The drawer style rack design allows easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up  Plate Capacity: 48 with lids 64 without lids 4×4 Configuration Dimensions (DxWxH): 14 5/8 x 5 7/16 x 9 3/4 inches (372 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) **Lock device is ordered separately.