Learn More
Alkali Scientific Upright Freezer Drawer Rack, Storage Drawers with Lids, 27 x 11 3/16 x 9 15/16 inches
SDP
Catalog No. 5000662522
Encompass_Preferred
Our freezer racks are constructed of high quality, corrosion resistant stainless steel designed for durability and long term laboratory use. These stainless steel freezer racks help organize samples while maximizing valuable freezer storage space. Handles at both ends accommodate freezers with right or left handed doors for convenient handling. The racks are designed for 96 Well Microtube Boxes, with each rack compartment holding a single 96 well microtube box and including a locking rod for secure storage. Our freezer racks are configured to fit all major ultra low ( 80°C) upright freezers on the market. Bin style organizer drawer freezer racks provide a systematic cold storage solution to accommodate tubes, bottles, boxes, vessels, and other lab containers. Ideal for cold rooms, 20°C freezers, and ultra low temperature freezers, these racks allow easy access to samples without removing the entire rack from the freezer, with clear wire mesh sides for quick v
Catalog No. 50-006-62522 Supplier Alkali Scientific Supplier No. UFTDLL11
Our freezer racks are constructed of high quality, corrosion resistant stainless steel designed for durability and long term laboratory use. These stainless steel freezer racks help organize samples while maximizing valuable freezer storage space. Handles at both ends accommodate freezers with right or left handed doors for convenient handling. The racks are designed for 96 Well Microtube Boxes, with each rack compartment holding a single 96 well microtube box and including a locking rod for secure storage. Our freezer racks are configured to fit all major ultra low ( 80°C) upright freezers on the market. Bin style organizer drawer freezer racks provide a systematic cold storage solution to accommodate tubes, bottles, boxes, vessels, and other lab containers. Ideal for cold rooms, 20°C freezers, and ultra low temperature freezers, these racks allow easy access to samples without removing the entire rack from the freezer, with clear wire mesh sides for quick v