Learn More
Thomas Scientific Drawer Freezer rack for 2" box, 5 x 5 cap
SDP
Catalog No. 50196056
Encompass_Preferred
Stainless Steel Freezer Drawer Rack for Upright Freezers for use with 2" or 3" Boxes
Catalog No. 50-196-056 Supplier Thomas Scientific Supplier No. 1201Y35
Stainless Steel Freezer Drawer Rack for Upright Freezers for use with 2" or 3" Boxes