Learn More
Labrepco Inc LABREPCO UFDMP-75 Upright Freezer Drawer Rack, Stainless Steel, 5 Drawers, Handles Included, 5 in
SDP
Catalog No. NC3404970
Encompass_Preferred
The LABREPCO INC UFDMP-75 Upright Freezer Drawer Rack offers a robust stainless steel solution for organizing samples in upright freezers. Its drawer design enhances accessibility while minimizing temperature fluctuations, protecting sensitive materials. Featuring five drawers with seven compartments each, integrated handles, and identification card holders, this rack optimizes vertical storage and improves sample traceability, making it an essential addition to any laboratory.
Catalog No. NC3404970 Supplier Labrepco Inc Supplier No. UFDMP75
The LABREPCO INC UFDMP-75 Upright Freezer Drawer Rack offers a robust stainless steel solution for organizing samples in upright freezers. Its drawer design enhances accessibility while minimizing temperature fluctuations, protecting sensitive materials. Featuring five drawers with seven compartments each, integrated handles, and identification card holders, this rack optimizes vertical storage and improves sample traceability, making it an essential addition to any laboratory.