Learn More
Labrepco Inc Stainless Steel Upright Freezer Drawer Rack for MATRIX Boxes, FluidX Boxes & Micronic Racks – 25 Box Capacity – 17.7″ (L) x 13.2″ (H)
SDP
Catalog No. NC1856480
Encompass_Preferred
Upright Freezer Drawer Racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks featuring handles, and identification card holders are made of corrosion resistant stainless steel. Drawer style rack design allows easy access to fewer racks at a time thus protecting other samples in the rack from warming up. Racks fit all standard types of upright freezers. Boxes not included.
Catalog No. NC1856480 Supplier Labrepco Inc Supplier No. UFDMX552
Upright Freezer Drawer Racks for 96 Well MATRIX boxes, FluidX Boxes & Micronic Racks featuring handles, and identification card holders are made of corrosion resistant stainless steel. Drawer style rack design allows easy access to fewer racks at a time thus protecting other samples in the rack from warming up. Racks fit all standard types of upright freezers. Boxes not included.