Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS
SDP
Catalog No. NC2175890
Encompass_Preferred
Upright Freezer Drawer Racks for Microtube Boxes (FluidX, Micronic Racks, etc.) – Rack Only – 6 Boxes Deep x 6 Boxes High
This item is not returnable. View return policy
Catalog No. NC2175890 Supplier Labrepco Inc Supplier No. UFDML66
Upright Freezer Drawer Racks for Microtube Boxes (FluidX, Micronic Racks, etc.) – Rack Only – 6 Boxes Deep x 6 Boxes High