Learn More
Thomas Scientific Drawer Freezer rack for 3" box, 3 x 2 cap
SDP
Catalog No. 50195978
Encompass_Preferred
Stainless Steel Freezer Drawer Rack for Upright Freezers for use with 2" or 3" Boxes
Catalog No. 50-195-978 Supplier Thomas Scientific Supplier No. 1201Y40
Stainless Steel Freezer Drawer Rack for Upright Freezers for use with 2" or 3" Boxes