Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Supplier:  Labrepco Inc UFDT062
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Blood Sample Tubes; Rack with 1 drawer; Capacity: 170 tubes of 4mL, 4.5mL, 6mL or 7mL; L x H x D: 419 x 228 x 140mm
Catalog No. 50-902-0252