Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Catalog No. 509020183
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; Crystal; Racks for standard 3 in. boxes; Capacity (Box): 12; L x H x D: 559 x 256 x 140mm
Catalog No. 50-902-0183 Supplier Labrepco Inc Supplier No. UFD433
Drawer Rack, Upright freezer; Labrepco; Crystal; Racks for standard 3 in. boxes; Capacity (Box): 12; L x H x D: 559 x 256 x 140mm