Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Supplier:  Labrepco Inc UFDLT101
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Blood Sample Tubes; Rack with 1 drawer; Capacity: 80 tubes of 8mL, 8.5mL or 10mL; L x H x D: 559 x 114 x 140mm
Catalog No. 50-902-0247