Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks forLarge 3.75″ Boxes- Rack Only- 5 Boxes Deep x 4 Boxes High
SDP
Catalog No. 503027521
Encompass_Preferred
For 3.75″ High Large Boxes These Upright Freezer Drawer Racks for Large 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Dimensions L x H x W: 30 1/4 x 16 1/16 x 5 15/16 (315.5 x 408 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-7521 Supplier Alkali Scientific Supplier No. UFR54L375
For 3.75″ High Large Boxes These Upright Freezer Drawer Racks for Large 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Dimensions L x H x W: 30 1/4 x 16 1/16 x 5 15/16 (315.5 x 408 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)