Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Supplier:  Labrepco Inc UFD342
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; Crystal; Racks for standard 2 in. boxes; Capacity (Box): 12; L x H x D: 419 x 240 x 140mm
Catalog No. 50-902-0105