Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Blood Sample Tubes, Fits 4ml, 4.5ml, 6ml, 7ml Tubes, 115 Tube Capacity, 1 Drawer
SDP
Catalog No. 503027956
Encompass_Preferred
For Blood Sample Tubes The UFDR-LT06-1 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 115 tubes for 4ml, 4.5ml, 6ml, 7ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 1/2 inches (564 x 140 x 114mm)
Catalog No. 50-302-7956 Supplier Alkali Scientific Supplier No. UFDRLT061
For Blood Sample Tubes The UFDR-LT06-1 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 115 tubes for 4ml, 4.5ml, 6ml, 7ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 1/2 inches (564 x 140 x 114mm)