Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Storage of Bottles, 2 Drawers, Area Capacity: 15″ x 4 1/2″
SDP
Catalog No. 503027963
Encompass_Preferred
For Storage Bottles Upright Freezer Drawer Rack for storage bottles features 2 drawers and is constructed of corrosion resistant stainless steel. Drawer racks facilitate both access to and storage of bottles without having to remove the whole rack from the freezer. These freezer racks for storage bottles include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Dimensions L x H x W : 16 11/16  x 9 15/16  x 5 1/2 inch (424 x 252 x 140mm)Capacity: Drawer Area 15 x 4 1/2 (Per Drawer) 383 x 114mm *Lock device is ordered separately.
Catalog No. 50-302-7963 Supplier Alkali Scientific Supplier No. UFDRB2
For Storage Bottles Upright Freezer Drawer Rack for storage bottles features 2 drawers and is constructed of corrosion resistant stainless steel. Drawer racks facilitate both access to and storage of bottles without having to remove the whole rack from the freezer. These freezer racks for storage bottles include handles and identification card holders. Security lock devices are also available for each rack though ordered separately. Dimensions L x H x W : 16 11/16  x 9 15/16  x 5 1/2 inch (424 x 252 x 140mm)Capacity: Drawer Area 15 x 4 1/2 (Per Drawer) 383 x 114mm *Lock device is ordered separately.