Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Catalog No. 509020249
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Blood Sample Tubes; Rack with 1 drawer; Capacity: 160 tubes of 8mL, 8.5mL or 10mL; L x H x D: 559 x 228 x 140mm
Catalog No. 50-902-0249 Supplier Labrepco Inc Supplier No. UFDLT102
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Blood Sample Tubes; Rack with 1 drawer; Capacity: 160 tubes of 8mL, 8.5mL or 10mL; L x H x D: 559 x 228 x 140mm