Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Standard Large 3.75″ Boxes- Rack Only- 5 Boxes Deep x 2 Boxes High
SDP
Catalog No. 503028104
Encompass_Preferred
For Large 3.75″ High Standard Boxes These Upright Freezer Drawer Racks for Large 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Dimensions L x H x W: 30 1/4 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-8104 Supplier Alkali Scientific Supplier No. UFR52L375
For Large 3.75″ High Standard Boxes These Upright Freezer Drawer Racks for Large 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Dimensions L x H x W: 30 1/4 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)