Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Blood Sample Tubes, Fits 8ml, 8.5ml, 10ml Tubes, 80 Tube Capacity, 1 Drawer
SDP
Catalog No. 503028117
Encompass_Preferred
For Blood Sample Tubes The UFDR-LT10-1 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 80 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 1/2 inches (564 x 140 x 114mm)
Catalog No. 50-302-8117 Supplier Alkali Scientific Supplier No. UFDRLT101
For Blood Sample Tubes The UFDR-LT10-1 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 80 tubes for 8ml, 8.5ml, 10ml sizes. This freezer rack features one drawer and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 4 1/2 inches (564 x 140 x 114mm)