Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Supplier:  Labrepco Inc UFDXLT152
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Centrifuge Tubes and Storage Bottles; Capacity: 208 Tubes (15mL); L x H x D: 680 x 252 x 140mm
Catalog No. 50-902-0229