Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Blood Sample Tubes, Fits 3ml Tubes, 250 Tube Capacity, 2 Drawers
SDP
Catalog No. 503027782
Encompass_Preferred
For Blood Sample Tubes The UFDR-LT03-2 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 250 tubes for 3ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 6 1/16 inches (564 x 140 x 154mm)
Catalog No. 50-302-7782 Supplier Alkali Scientific Supplier No. UFDRLT032
For Blood Sample Tubes The UFDR-LT03-2 Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 250 tubes for 3ml sizes. This freezer rack features two drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 6 1/16 inches (564 x 140 x 154mm)