Learn More
Thomas Scientific Drawer Rack 100cell hinged box 3 x 3
SDP
Catalog No. 50196024
Encompass
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-024 Supplier Thomas Scientific Supplier No. 1149N91
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.