Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Supplier:  Labrepco Inc UFDMP44
Encompass_Preferred
Upright Freezer Drawer Racks for 96- & 384-Well Microtiter Plates, 96-Well Microtube Boxes and Micronic ROBO Racks. Dimensions (LxHxD): 14-1/2 in. x 9-11/16 in. x 5-7/16 in.. Capacity: 48 plates with lids/ 16 Microtube Boxes/ 16 Micronic ROBO Racks.
Catalog No. 50-902-0272