Learn More
Labrepco Inc UPRIGHT FREEZER DRAWER RACKS F
SDP
Catalog No. 509020230
Encompass_Preferred
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Centrifuge Tubes and Storage Bottles; Capacity: 30 Tubes (50mL); L x H x D: 419 x 126 x 140mm
Catalog No. 50-902-0230 Supplier Labrepco Inc Supplier No. UFDT501
Drawer Rack, Upright freezer; Labrepco; KSE Scientific; For Centrifuge Tubes and Storage Bottles; Capacity: 30 Tubes (50mL); L x H x D: 419 x 126 x 140mm