Learn More
Thomas Scientific Rack with 3 drawers (4 compartments per drawer) for Microtiter Plates
SDP
Catalog No. 50195999
Encompass_Preferred
Racks are for 96-well & 384-well Microtiter Plates. Drawer Racks allow an easy access to fewer Microtiter plates at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-195-999 Supplier Thomas Scientific Supplier No. 1148X47
Racks are for 96-well & 384-well Microtiter Plates. Drawer Racks allow an easy access to fewer Microtiter plates at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.