Learn More
Thomas Scientific Upright Freezer Drawer Racks for 12 x 100 Place Hinged Top Boxes, Holds 12 Boxes (4x3), 17 1/4"L x 9 11/16"H x 6 1/2"D
SDP
Catalog No. 50196041
Encompass_Preferred
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-041 Supplier Thomas Scientific Supplier No. 1148X15
Add to Cart
Add to Cart
Racks are for 100-Cell Hinged Top Plastic Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.