Learn More
Thomas Scientific Upright Freezer Drawer Racks for 96 Well Deep Well Plates, Rack w/5 Drawers (7 Compartments per Drawer), Holds 35 Plates. 25"L x 10 1/8"H x 5 1/2"D.
SDP
Catalog No. 50196057
Encompass_Preferred
Racks are for 96 Deep-Well Microtiter Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-057 Supplier Thomas Scientific Supplier No. 1148W96
Racks are for 96 Deep-Well Microtiter Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.