Learn More
Alkali Scientific Upright Freezer Rack for 100 Place Slide Boxes, 3 x 8 Configuration, 24 Box Capacity
SDP
Catalog No. 5000662811
Encompass_Preferred
For 100 Place Slide Boxes The UFDS 100 38 drawer style upright freezer racks for storage of 100 place slide boxes, features a 3 boxes deep by 8 boxes high configuration for a total storage of 24 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. Drawer freezer racks also feature a label card holder on the front of every rack for easy identification and organization of racks and samples. This freezer rack does not come standard with 100 place slide boxes. Dimensions L x H x W : 22 3/16 x 13 13/16 x 8 5/8 inch (563.5 x 351 x 219 mm) Compartment Dimensions l x h x w: 7 1/8 x 1 1/2 x 8 1/4 inch (180.5 x 38.4 x 210 mm) *Security lock device is ordered separately.
Catalog No. 50-006-62811 Supplier Alkali Scientific Supplier No. UFDS10038
For 100 Place Slide Boxes The UFDS 100 38 drawer style upright freezer racks for storage of 100 place slide boxes, features a 3 boxes deep by 8 boxes high configuration for a total storage of 24 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. Drawer freezer racks also feature a label card holder on the front of every rack for easy identification and organization of racks and samples. This freezer rack does not come standard with 100 place slide boxes. Dimensions L x H x W : 22 3/16 x 13 13/16 x 8 5/8 inch (563.5 x 351 x 219 mm) Compartment Dimensions l x h x w: 7 1/8 x 1 1/2 x 8 1/4 inch (180.5 x 38.4 x 210 mm) *Security lock device is ordered separately.