Learn More
Research Products International Corp Upright Freezer Rack for 96-Deep Well Plates, Rack Dimensions (Inches): 18 1/2 x 5 1/2 x 9 3/16H
SDP
Catalog No. 501977620
Encompass
Fits micro-plates with dimensions of 5'' x 3-3/8'' x 1-5/8'' H
Catalog No. 50-197-7620 Supplier Research Products International Corp Supplier No. 247556
Fits micro-plates with dimensions of 5'' x 3-3/8'' x 1-5/8'' H