Learn More
Alkali Scientific Upright Freezer Rack for 25 Place Slide Boxes, 6 x 6 Configuration, 36 Box Capacity
SDP
Catalog No. 5000662525
Encompass_Preferred
For 25 Place Slide Boxes Our Freezer Storage Racks are constructed of high quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our freezer racks have been configured to fit all major ultra low ( 80°) freezers on the market; racks are designed for ultra low upright freezers. Racks are for 25 Place Slide Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel. Dimensions L x H x W: 22 9/16 x 10 3/16 x 6 inch (572.5 x 259 x 152 mm) Compartment Dimensions l x h x w: 3 11/16 x 1 1/2 x 5 9/16 inch (92.5 x 38 x 141mm) *Security lock device is ordered separately.
Catalog No. 50-006-62525 Supplier Alkali Scientific Supplier No. UFDS2566
For 25 Place Slide Boxes Our Freezer Storage Racks are constructed of high quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our freezer racks have been configured to fit all major ultra low ( 80°) freezers on the market; racks are designed for ultra low upright freezers. Racks are for 25 Place Slide Boxes. Drawer Racks allow an easy access to fewer microtube boxes at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel. Dimensions L x H x W: 22 9/16 x 10 3/16 x 6 inch (572.5 x 259 x 152 mm) Compartment Dimensions l x h x w: 3 11/16 x 1 1/2 x 5 9/16 inch (92.5 x 38 x 141mm) *Security lock device is ordered separately.