Learn More
Research Products International Corp Upright Freezer Rack for Plastic 2 Inch Boxes, 4 x 4 Array, 5 1/2 x 22 1/2 x 9 1/4 Inches
SDP
Catalog No. 50998178
Encompass
Upright freezer rack for storage of 0.5 ml, 50-cell plastic microtube boxes. Racks are in a 4 x 4 array.
Catalog No. 50-998-178 Supplier Research Products International Corp Supplier No. 247599
Upright freezer rack for storage of 0.5 ml, 50-cell plastic microtube boxes. Racks are in a 4 x 4 array.