Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Stainless Access Freezer Rack, Holds (8) 3.75" Boxes, Rack Dimensions 22 1/8L x 8 1/16H x 5 1/2W" (562 x 205 x 140mm) Compartment Dimensions 5 3/8L x 3 15/16H x 5 3/8W" (136 x 100 x 137mm) 4x2
Catalog No. NC2208081
Encompass_Preferred
Rack Designed to hold (8) Standard 3.75H"Cardboard Boxes, and Plastic (Polypropylene or Polycarbonate) Boxes, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208081 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UF42375
Rack Designed to hold (8) Standard 3.75H"Cardboard Boxes, and Plastic (Polypropylene or Polycarbonate) Boxes, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.