Learn More
Labrepco Inc UPRIGHT FREEZER RACK, 4 DRAWER
SDP
Catalog No. NC2165463
Encompass_Preferred
Upright Freezer Racks with 4 Drawers (7 Compartments Each) for 96 Deep- Well Microtiter Plates and Micronic LOBO Racks – Rack Only – 7 Plates Deep x 4 Plates High
Catalog No. NC2165463 Supplier Labrepco Inc Supplier No. UFDDP74
Upright Freezer Racks with 4 Drawers (7 Compartments Each) for 96 Deep- Well Microtiter Plates and Micronic LOBO Racks – Rack Only – 7 Plates Deep x 4 Plates High