Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Side Access Freezer Rack, Holds (30) 96-Well Microtube Boxes,Rack Dims 22 1/2L x 11 3/16H x 5 1/2W" (571 x 284 x 140mm) Compartment Dims 3 9/16L x 2 3/16H x 5 3/16W" (91 x 55 x 131mm) 6x5
Catalog No. NC2208103
Encompass_Preferred
Rack Designed to hold (30) 96-Well Microtube Boxes (2 1/8"H x5W x3 3/8L" (54 x 127 x 85.7mm), Locking Rods Included, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208103 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFTB65L
Rack Designed to hold (30) 96-Well Microtube Boxes (2 1/8"H x5W x3 3/8L" (54 x 127 x 85.7mm), Locking Rods Included, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.