Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack, Bin Style Organizer, with Lid 12 x 11 Bin, 16 9/16 x 11 x 12 13/16"
SDP
Catalog No. 503028403
Encompass_Preferred
One Bins Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 16 9/16 x 11 x 12 13/16 inches (421 x 279 x 325mm)Compartment Dimensions  l x h x w: 16 1/8 x 12 11/16 x 10 11/16 inch (409 x 323 x 272mm)
Catalog No. 50-302-8403 Supplier Alkali Scientific Supplier No. UFTS1211
One Bins Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 16 9/16 x 11 x 12 13/16 inches (421 x 279 x 325mm)Compartment Dimensions  l x h x w: 16 1/8 x 12 11/16 x 10 11/16 inch (409 x 323 x 272mm)