Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack, Bin Style Organizer, with Lid 6 x 11 Bin, 22 1/16 x 11 x 6 5/8"
SDP
Catalog No. 503027770
Encompass_Preferred
For Mixed Use Storage Bins Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 22 1/16 x 11 x 6 5/8 inches (560 x 279 x 168mm)Compartment Dimensions  l x h x w: 21 9/16 x 6 9/16 x 10 11/16 inch (548 x 166 x 272mm)
Catalog No. 50-302-7770 Supplier Alkali Scientific Supplier No. UFTM0611
For Mixed Use Storage Bins Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 22 1/16 x 11 x 6 5/8 inches (560 x 279 x 168mm)Compartment Dimensions  l x h x w: 21 9/16 x 6 9/16 x 10 11/16 inch (548 x 166 x 272mm)