Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack, Bin Style Organizer, with Lid 4 x 5 Bin, 16 1/3 x 4 4/5 x 5 3/7"
SDP
Catalog No. 503027771
Encompass_Preferred
For Mixed Use Storage Bins Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions L x H x W: 16 9/16 x 4 3/4 x 5 7/16 inch ( 421 x 120 x 138mm)Compartment Dimensions l x h x w: 16 1/8 x 4 3/4 x 5 3/16inch (409 x 120 x 132mm)
Catalog No. 50-302-7771 Supplier Alkali Scientific Supplier No. UFTS045
For Mixed Use Storage Bins Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions L x H x W: 16 9/16 x 4 3/4 x 5 7/16 inch ( 421 x 120 x 138mm)Compartment Dimensions l x h x w: 16 1/8 x 4 3/4 x 5 3/16inch (409 x 120 x 132mm)