Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack, Bin Style Organizer, with Lid 11x11 Bin, 26 3/4 x 11 x 11"
SDP
Catalog No. 503028166
Encompass_Preferred
For Mixed Use Storage Bins Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 26 3/4 x 11 x 11 inches (680 x 279 x 280mm)Compartment Dimensions  l x h x w: 26 5/16  x 10 15/16 x 10 11/16 inch (668 x 278 x 272mm)
Catalog No. 50-302-8166 Supplier Alkali Scientific Supplier No. UFTL1111
For Mixed Use Storage Bins Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 26 3/4 x 11 x 11 inches (680 x 279 x 280mm)Compartment Dimensions  l x h x w: 26 5/16  x 10 15/16 x 10 11/16 inch (668 x 278 x 272mm)