Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack, Bin Style Organizer, with Lid 8x5" Bin, 26 3/4 x 5 7/16 x 8 7/8"
SDP
Catalog No. 503026699
Encompass_Preferred
1 Bin Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 26 3/4 x 5 7/16 x 8 7/8 inches (680 x 138 x 225mm)Compartment Dimensions: 26 1/4 x 5 1/4 x 8 7/8 inches (665 x 134 x 225mm)
Catalog No. 50-302-6699 Supplier Alkali Scientific Supplier No. UFTL085
1 Bin Our Freezer Racks are constructed of high-quality, corrosion resistant stainless steel.  Bin Style Organizer Freezer Racks provide a systematic cold storage solution to accommodate nearly any tube, bottle, box, vessel, and more. These freezer racks are perfect for cold rooms, -20C freezers and ultra-low temp freezers, and come complete with lids for added security and see through, wire mesh, sides for easy accessibility.  Dimensions (DxWxH): 26 3/4 x 5 7/16 x 8 7/8 inches (680 x 138 x 225mm)Compartment Dimensions: 26 1/4 x 5 1/4 x 8 7/8 inches (665 x 134 x 225mm)