Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Rack, Holds (8) Centrifuge Boxes, Rack Dimensions 24 13/16L x 10 3/16H x 5 7/8W" (631 x 259 x 149mm) with Compartment Dimensions 6L x 5 1/16H x 5 13/16W" (152 x 128 x 147.5mm) 4 x 8
Catalog No. NC2208042
Encompass_Preferred
Rack Designed to hold (8) 5"H 15ml and 50ml Centrifuge Tube Boxes, (4 15/16H x 5 13/16L x 5 13/16W" (125 x 148 x 148mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208042 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFLB42
Rack Designed to hold (8) 5"H 15ml and 50ml Centrifuge Tube Boxes, (4 15/16H x 5 13/16L x 5 13/16W" (125 x 148 x 148mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.