Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 15 ml & 50 ml Tube Boxes | Rack Only | 4 Boxes Deep x 2 Boxes High
SDP
Catalog No. 503029352
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 25 with lids 30 without lids The stainless steel upright freezer rack for storage of 15ml & 50ml Centrifuge Tube Boxes is designed to fit a total of 8 boxes. These freezer racks come standard with Locking Rods and are constructed of corrosion resistant stainless steel. Security lock devices & storage boxes are also available for each rack though ordered separately. *Tubes and boxes not included Dimensions (DxWxH): 3 5/8 x 5 1/2 x 16 13/16 inches (92 x 140 x 427mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.
Catalog No. 50-302-9352 Supplier Alkali Scientific Supplier No. CFMP5
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 25 with lids 30 without lids The stainless steel upright freezer rack for storage of 15ml & 50ml Centrifuge Tube Boxes is designed to fit a total of 8 boxes. These freezer racks come standard with Locking Rods and are constructed of corrosion resistant stainless steel. Security lock devices & storage boxes are also available for each rack though ordered separately. *Tubes and boxes not included Dimensions (DxWxH): 3 5/8 x 5 1/2 x 16 13/16 inches (92 x 140 x 427mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.