Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 2″ Cardboard Box; Made to fit Panasonic Freezers; Rack Only; 5x6 Configuration, 30 Box Capacity
SDP
Catalog No. 503029046
Encompass_Preferred
Panasonic Specific Model: Upright freezer racks for storage of 2-inch boxes, features a 5 boxes deep by 6 boxes high configuration for a total storage of 30 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. This drawer style freezer rack does not come standard with storage boxes. Dimensions (D x H x W): 27  x 14 1/8 x 5 1/2 inches (687 x 321 x 140mm)Compartment Dimensions: 5 3/8  x 2 1/8 x 5 1/4inches (137.4 x 54.4 x 133mm)
Catalog No. 50-302-9046 Supplier Alkali Scientific Supplier No. UFDR562S
Panasonic Specific Model: Upright freezer racks for storage of 2-inch boxes, features a 5 boxes deep by 6 boxes high configuration for a total storage of 30 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. This drawer style freezer rack does not come standard with storage boxes. Dimensions (D x H x W): 27  x 14 1/8 x 5 1/2 inches (687 x 321 x 140mm)Compartment Dimensions: 5 3/8  x 2 1/8 x 5 1/4inches (137.4 x 54.4 x 133mm)