Learn More
Alkali Scientific Stainless Steel Freezer Racks for 3" Boxes, 4 x 3 Configuration, 12 Box Capacity
SDP
Catalog No. 502389065
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks fit standard 3" cardboard, plastic, or metal boxes. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 9 3/8 inches (564 x 140 x 238mm). Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (137 x 140 x 78mm).
Catalog No. 50-238-9065 Supplier Alkali Scientific Supplier No. UFR433
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks fit standard 3" cardboard, plastic, or metal boxes. Dimensions (DxWxH): 22 3/16 x 5 1/2 x 9 3/8 inches (564 x 140 x 238mm). Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (137 x 140 x 78mm).